Daptomycin ≥93% (by HPLC)

Supplier: Enzo Life Sciences

BML-A201-0100 BML-A201-0020
76002-358EA 143.79 USD
76002-358 76002-360
Daptomycin ≥93% (by HPLC)
Daptomycin

Lipopeptide antibiotic


Novel lipopeptide antibiotic active against Gram positive bacteria.

Formula: C₇₂H₁₀₁N₁₇O₂₆
MDL Number: MFCD08282794
CAS Number: 103060-53-3
https://us.vwr.com/stibo/web/std.lang.all/34/29/45403429.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR