Necrostatin-7 ≥98% (by HPLC, TLC)

Supplier: Enzo Life Sciences

430-170-M005
76002-024EA 138.18 USD
76002-024
Necrostatin-7 ≥98% (by HPLC, TLC)
Necrostatin-7

Necroptosis inhibitor


Necroptosis inhibitor. The biological activity of necrostatin-7 (Nec-7) is different from that of necrostatin-1 to -5 (Nec-1 to -5), as Nec-7 does not inhibit RIP1 kinase. Nec-7 may target an additional regulatory molecule in the pathway.

Formula: C₁₆H₁₀FN₅OS₂
CAS Number: 351062-08-3
https://us.vwr.com/stibo/web/std.lang.all/07/18/45390718.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR