Phomopsin A ≥98% (by HPLC)

Supplier: Enzo Life Sciences

350-417-M001
76002-194EA 721.28 USD
76002-194
Phomopsin A ≥98% (by HPLC)
Phomopsin A

Isolated from Phomopsis leptostromiformis MST-FP2068.


Isolated from Phomopsis leptostromiformis MST-FP2068.

A potent mycotoxin and anti-mitotic compound. Causes lupinosis in livestock fed infected lupins. Binds selectively to dimeric tubulin, inhibiting the formation of the microtubule spindle to block cell division. Binds to tubulin at a site overlapping that of vinblastine (Prod. No. ALX-350-257) and maytansine. Protects tubulin from decay.


A potent mycotoxin and anti-mitotic compound. Causes lupinosis in livestock fed infected lupins. Binds selectively to dimeric tubulin, inhibiting the formation of the microtubule spindle to block cell division. Binds to tubulin at a site overlapping that of vinblastine (Prod. No. ALX-350-257) and maytansine. Protects tubulin from decay.

Formula: C₃₆H₄₅ClN₆O₁₂
MW: 789.2 g/mol
CAS Number: 64925-80-0
https://us.vwr.com/stibo/web/std.lang.all/03/88/45390388.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR