ASSA350-415
350-415-M001 350-415-MC25
76002-006EA 78.4 USD
76002-006 76002-008
ASSA350-415
Free radical scavenger.
Antioxidant, with a radical scavenging activity toward DPPH and superoxide.
Soluble in DMSO or hot methanol; insoluble in hexane.