ADIPSYN3010
SYN-3010-M050 SYN-3010-M010 SYN-3010-M005 SYN-3010-M001
102992-888EA 246.46 USD
102992-888 102992-886 102992-884 102992-882
ADIPSYN3010
Azithromycin is an antibiotic for inhibition of parasite growth.
  • Antibiotic
  • Inhibition of parasite growth
Azithromycin is an antibiotic for inhibition of parasite growth with an IC(50) of 8.4µM.
Solid
Soluble in DMSO or ethanol