Tetracycline hydrochloride (synthetic) ≥99% (by HPLC)

Supplier: ADIPOGEN CORP MS

CDX-T0096-G005 CDX-T0096-G025
102991-118EA 74 USD
102991-118 102991-116
Tetracycline hydrochloride (synthetic) ≥99% (by HPLC)
Tetracycline hydrochloride

Tetracycline is a broad-spectrum antibiotic that prevents bacterial growth by inhibiting protein synthesis.


  • Selective agent in cell culture systems
  • Prevents bacterial growth by inhibiting protein synthesis
  • Binds to a single site in the 30S ribosomal subunit


Tetracycline is a broad-spectrum antibiotic that prevents bacterial growth by inhibiting protein synthesis. It binds to a single site in the 30S ribosomal subunit which prevents attachment of aminoacyl tRNA to the ribosomal acceptor site. It is used in cell biology as a selective agent in cell culture systems. Tetracycline is toxic to prokaryotic and eukaryotic cells and selects for cells harboring the bacterial tetR gene, which are resistant to the antibiotic.


Yellow to orange powder


Soluble in water (10 mg/mL), DMSO, methanol or 0.1 N HCl (10 mg/mL)

Formula: C₂₂H₂₅ClN₂O₈
MW: 480.9 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00078142
CAS Number: 64-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR