Flumequine (synthetic) ≥97% (by HPLC)
Supplier: ADIPOGEN CORP MS
Synonyms:
9-Fluoro-5-methyl-1-oxo-6,7-dihydro-1H,5H-pyrido[3,2,1-ij]quinoline-2-carboxylic acid
, 9-Fluoro-1,5,6,7-tetrahydro-5-methyl-1-oxopyrido[3,2,1-ij]quinoline-2-carboxylic acid
Flumequine is a fluoroquinolone synthetic chemotherapeutic antibiotic used to treat bacterial infections.
- Inhibits topoisomerases
- Used to study hepatocarcinogenicity
- Targets primarily gram-negative bacteria
Targets primarily gram negative bacteria, especially those which cause enteric infections in animals. It is used to study processes that affect mammalian chromosome and DNA unwinding at the level of gyrase/topoisomerases. It inhibits topoisomerases, which are needed for the transcription and replication of bacterial DNA. The inhibition of the topoisomerases results in strand breakage of the bacterial chromosome, supercoiling and resealing. Therefore, DNA replication and transcription is inhibited. It was also used to study hepatocarcinogenicity and DNA damage in mice and the mechanisms of quinolone resistance.
White to off-white powder
Soluble in toluene
Formula:
C₁₄H₁₂FNO₃ MW: 261.25 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00079298 CAS Number: 42835-25-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...