Would you like to log in again?
Mycotoxin TargoSet™, AdipoGen Life Sciences
Supplier: ADIPOGEN CORP MS
The Mycotoxin TargoSet™ offers a panel of well-characterized and highly pure compounds for screening experiments, cell-based research or as standard compounds.
  • Used in research as antibiotics
  • Screening experiments
  • Wide spectrum of biological and pharmacological activities
Mycotoxins are secondary metabolites produced by microfungi. Because of their wide spectrum of biological and pharmacological activities, some are used in research as antibiotics, cell growth promotants or cytotoxic compounds in cancer cells. Mycotoxins are harmful to animal and human health if they enter the food chain via contaminated crops and fruits or through processed food and feed products.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Description Size Packaging Supplier No. VWR Catalog Number Unit Price Quantity
Mycotoxin TargoSet™ 1 Set Plastic Box BVT-L002-1 102981-226 Each Retrieving Restricted
Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFICATIONS
Storage Conditions Freezer
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty