Venturicidin A (from Streptomyces aureofaciens DSM 40731) ≥98% (by 1H-NMR, HPLC)

Supplier: ADIPOGEN CORP MS

BVT-0454-M001
102989-068EA 166.15 USD
102989-068
Venturicidin A (from Streptomyces aureofaciens DSM 40731) ≥98% (by 1H-NMR, HPLC)
Venturicidin A

Venturicidin A is a 20-Membered macrolide glycoside.


  • Antibiotic and anti-fungal agent
  • Shows anti-malarial activity
  • Inhibitor of F0F1-ATPase


Shows anti-trypanosomal and anti-malarial activity. Potent inhibitor of mitochondrial and bacterial ATP synthase. Na+-translocating ATP synthases inhibitor.


White solid


Soluble in DMSO, methanol, acetone or dichloromethane. Insoluble in hexane or water

Storage Temperature: Refrigerator
CAS Number: 33538-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR