Skyrin (from Penicillium species) ≥98% (by HPLC)
Supplier: ADIPOGEN CORP MS
Synonyms:
Endothianin
, Rhodophyscin
BVT-0445-M005
102989-042EA
623.08
USD
102989-042
Skyrin (from Penicillium species) ≥98% (by HPLC)
Skyrin
Skyrin is a non-peptidic, anti-diabetic agent.
- Cytotoxic
- Non-peptidic
- Antioxidant (free radical scavenger)
Mycotoxin. Antibiotic (incl. MRSA). Non-peptidic anti-diabetic agent (receptor-selective glucagon antagonist). Inhibitor of botulinum neurotoxin serotype A (BoNTA).
Dark Red solid
Soluble in DMSO
Formula:
C₃₀H₁₈O₁₀ MW: 538.46 Storage Temperature: Freezer |
CAS Number:
602-06-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...