ADIPBVT0418
BVT-0418-M005 BVT-0418-M001
102988-974EA 131.54 USD
102988-974 102988-976
ADIPBVT0418
Sceliphrolactam is an unusual polyene antibiotic.
  • Polyene antibiotic
  • Anti-fungal, anti-bacterial
  • Inhibits amphothericin B-resistant Candida albicans
Anti-fungal and anti-bacterial agent. Inhibits amphothericin B-resistant Candida albicans (MIC 4 µg/mL).
Yellow powder
Soluble in DMSO or pyridine. Slightly soluble in methanol or acetone