β-Naphthocyclinone (from Streptomyces arenae) ≥98% (by HPLC, NMR)

Supplier: ADIPOGEN CORP MS

BVT-0250-M001
102988-908EA 166.15 USD
102988-908
β-Naphthocyclinone (from Streptomyces arenae) ≥98% (by HPLC, NMR)
beta-Naphthocyclinone

beta-Naphthocyclinone is an Isochromane quinone antibiotic.


  • Anti-bacterial
  • Isochromane quinone antibiotic


Isochromane quinone antibiotic. Anti-bacterial.


Red solid


Soluble in DMSO, methanol, acetone or chloroform

Storage Temperature: Refrigerator
CAS Number: 55050-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR