Palmarumycin C3
Supplier: ADIPOGEN CORP MS
BVT-0066-M001
102981-194EA
180
USD
102981-194
Palmarumycin C3
Palmarumycin C3
Palmarumycin C3-5,8-quinone is an oxidation product of palmarumycin C3.
- Spirobisnaphthalene derivative
- Oxidation product
Spirobisnaphthalene derivative. Oxidation product of palmarumycin C3. This compound has not yet been thoroughly tested for its biological activity.
Formula:
C₂₀H₁₂O₆ |
CAS Number:
159934-11-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...