Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Vaplock™ GL38 Waste Caps and Waste Kits, Western Analytical Products

Supplier: WESTERN ANALYTICAL PRODUCTS MS
WAK38P-10 WM38P-10 WM38P-4 WM38X-10 WM38X-4 WAK38X-10
Vaplock™ closed system design reduces chemical evaporation and exposure to hazardous vapors.

  • Minimize exposure to harmful chemical vapors
  • Collect solvent waste from multiple effluent streams
  • Highly adaptable and conforms to a broad range of containment types

Featuring a vapor tight connection, waste manifolds are designed to fit standard GL38 threaded lab bottles. Chemical-resistant materials of construction include polypropylene or PTFE manifold and insert, with polypropylene spin collar. These versatile manifolds can be configured for barbed connections, compression fittings, 1/4-28 fittings, and ferrule sets. All manifolds are designed to accommodate the EF-100 Filter and indicator.

Ordering information: Kits include (1) 10-port waste manifold, (3)1/4" NPT polypropylene plugs, (6) 1/4-28 PTFE plugs, (1)1/2" NPT polypropylene plug, (1) organic vapor filter, (2) 2.2 mm O.D. hard wall tubing connections, (1) 2.5 mm O.D. hard wall tubing connection, (3) 1/8” O.D. hard wall tubing connections, (2) 1/16” O.D. hard wall tubing connections, (3) barbed tubing connection 1/4" I.D. tubing x 1/4" NPT polypropylene, (1) barbed tubing connection 1/8" I.D. tubing x 1/4" NPT polypropylene, (1) barbed tubing connection 3/16" I.D. tubing x 1/4" NPT polypropylene, (1) barbed tubing connection 3/8" I.D. tubing x 1/4" NPT polypropylene, (1) barb to barb Y connector with 1/4" (6.25 mm) barbs, polypropylene, (1) hard wall tube extension with union connections and 5' of 1/8" O.D. PTFE tubing, (1) filter adapter 1/4" NPT x 1/2" NPT polypropylene, and (1) breakthrough color indicator for Vaplock™ closed system organic vapor filter.

Container Not Included.



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All

Waste Manifold Kits
Material Port Size Supplier No. VWR Catalog Number Unit Price Quantity
PP Manifold (1) 1/2 NPT, (3) 1/4 NPT, and (6) 1/4-28 Ports WAK38P-10 103384-360 Kit of 1 Retrieving
PTFE Manifold (1) 1/2 NPT, (3) 1/4 NPT, and (6) 1/4-28 Ports WAK38X-10 103384-362 Kit of 1 Retrieving

Waste Manifold Only
Material Port Size Supplier No. VWR Catalog Number Unit Price Quantity
PP Manifold (1) 1/2 NPT, (3) 1/4 NPT, and (6) 1/4-28 Ports WM38P-10 103384-364 Each Retrieving
PP Manifold (3) 1/4 NPT and (1) 1/2 NPT Ports WM38P-4 103384-366 Each Retrieving
PTFE Manifold (1) 1/2 NPT, (3) 1/4 NPT, and (6) 1/4-28 Ports WM38X-10 103384-368 Each Retrieving
PTFE Manifold (3) 1/4 NPT and (1) 1/2 NPT Ports WM38X-4 103384-370 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty