Fletcher's Media, Hardy Diagnostics
X67
75993-566PK 167.99 USD
75993-566
Fletcher's Media, Hardy Diagnostics
Hardy Diagnostics Fletcher's Media is an enriched semi-solid medium recommended for the cultivation of Leptospira spp.