Would you like to log in again?
Vaplock™ GL25 Supply Caps, Western Analytical Products
Supplier: WESTERN ANALYTICAL PRODUCTS MS
The Vaplock™ closed system design reduces chemical evaporation and exposure to hazardous vapors.
  • Closed system design reduce exposure to hazardous chemical vapors
  • Anti-Twist design prevents twisting of tubing when the cap is attached or removed
  • Chemical compatibility with laboratory solvents
Closed system design reduces chemical evaporation and exposure to hazardous vapors with Vaplock™ closed system solvent reservoir caps. Combine with secure fluid connections and Vaplock air inlet or relief valves to maintain pressure equilibrium within the bottle.
Durable port threads are designed to accept industry-standard 1/4-28 threaded fittings for use with semi-rigid tubing equal to or less than 1/8” (3.2 mm) outside diameter.
The Anti-Twist design prevents twisting of tubing when the cap is removed or attached, as the inner body rotates independently of the outer collar.
Chemically compatible caps are constructed of inert PTFE. The body of the cap provides ideal chemical compatibility with laboratory solvents. Under normal use, the outer polypropylene collar is non-wetted. Caps may be autoclaved to 140°C.
Please inquire for other material options and custom designs.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Description Material Port Size Color Supplier No. VWR Catalog Number Unit Price Quantity
GL25 Supply Caps with Standard Threaded Ports PTFE with PP Spin Collar (2) 1/4-28 Ports Blue S25X-2D 103384-356 Each Retrieving
GL25 Supply Caps with Standard Threaded Ports PTFE with PP Spin Collar (3) 1/4-28 Ports Blue S25X-3D 103384-358 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty