Brady® BMP®61 Label Printer, USB and USB/WiFi Capable, Brady Worldwide
146229 146226
75840-030EA 1052.35 USD
75840-030 75840-024
Brady® BMP®61 Label Printer, USB and USB/WiFi Capable, Brady Worldwide
The BMP®61 label printer is designed for efficient identification of wires, cables, and components
  • The BMP®61 Label Printer is a rugged and reliable partner in the field, and a dependable workhorse in the shop
  • Wide cover opening and large material bay offers up to 2" wide, easy material loading and bulk size rolls
  • With a USB 2.0 port and a USB flash drive, users can import and export printer generated label files, lists, and graphics to a BMP®61 Printer
  • Simple user interface, color touch screen, and QWERTY style keypad offer easy and quick navigation
Includes hard case, NiMH rechargeable battery, power cord, AC adapter/charger, PTL-19-423, USB cord, M61-R6010 ribbon, utility strap, product CD, printed Quick Start Guide, and Driver CD.
It can handle tough industrial applications with its reliable materials.
Certifications: FCC Part 15 Class A.
Accessories information: Plastic battery cover snaps in place over the battery compartment and ensures batteries remain securely in place.