Brady® BMP®21 LAB Printer Kit with AC Adapter, Brady Worldwide
139817
10117-450EA 475.9 USD
10117-450
Brady® BMP®21 LAB Printer Kit with AC Adapter, Brady Worldwide
The BMP®21-LAB Printer can be used for slides, tubes, flasks, beakers, bottles, and much more in just a matter of seconds.
  • Drop-lock-and-print continuous materials up to 0.75" wide
Why label with a pen that can smear or fade when you can print crisp and reliable labels from the BMP®21-LAB label printer? Label slides, tubes, flasks, beakers, bottles, and much more in just a matter of seconds
Kit includes BMP®21-LAB Printer, rechargeable Li-ION battery, and an AC adaptor/battery charger. Kit also includes built-in templates for lab vials and tubes, laboratory and Greek symbols, and a standard two-year warranty.
The BMP®21-LAB Printer can take on any labeling project in the toughest lab environments, from autoclaves and liquid nitrogen to freezers and hot water baths.
Certifications: FCC Part 15 Class A.