Brady® Code Reader™ 3600, Brady Worldwide
Supplier: Brady
143565 143563 143566 143561 143564 143562
10796-266EA 1284.76 USD
10796-266 10796-264 10796-268 10796-270 10796-274 10796-272
Brady® Code Reader™ 3600, Brady Worldwide
Improve data collection operations with the Code Reader™ 3600.
Code Reader™ 3600 is an ultra-fast, durable barcode reader with an aggressive scan engine. Dual field optics make transitioning from decoding wide to small barcodes easy. Scanner packages are combined with cables, chargers, batteries, handles, and stands.