Non-Adhesive Polyester Printable Tags, B30, Brady
Supplier: Brady
142224 142223 142222 142226
10018-050EA 380.07 USD
10018-050 10018-046 10018-048 10018-044
Non-Adhesive Polyester Printable Tags, B30, Brady
Labels
Made of non-adhesive semi-rigid card stock.
Polyester tags can be used with label makers or printers to print small tags.
NOTE: BBP®31 printers must have version 001.010.003 or higher AND firmware 107 or higher in order to use this pre-print, pre-cut tag supply.
Die-cut rolls are pre-cut to a specific size. Tags are compatible with BBP®30, BBP®31, BBP®33, BBP®35, and BBP®37 devices.
Certifications: Conforms to OSHA Specification 1910.145.