Brady® Code Reader™ 1400, Brady Worldwide
133070
10796-254EA 559.6 USD
10796-254
Brady® Code Reader™ 1400, Brady Worldwide
The compact Code Reader™ 1400 Scanner improves data collection operations with its high performance scan engine, patented dual-field optical platform, and intuitive targeting.
  • High-speed, omnidirectional reading of 1D and 2D barcodes and glare reduction technology for reading barcodes on shiny surfaces
  • Features dual field optics with both high-density and wide field in the same unit, plus manual or automatic triggering
  • Offers data editing and parsing with JavaScript and disinfectant-ready housing