SP Wilmad-LabGlass CHROMWARE® THF-Resistant Three Valve Cap Reservoir Systems, SP Industries
BP-1931-020 BP-1931-001 BP-1931-010 BP-1931-002 BP-1931-005
14207-614EA 2269.77 USD
14207-614 14205-844 14207-472 14207-500 14206-156
SP Wilmad-LabGlass CHROMWARE® THF-Resistant Three Valve Cap Reservoir Systems, SP Industries
CHROMWARE® Three Valve Reservoir Systems are recommended for applications where on-off control of all three ports on the cap is necessary.
For example, HPLC mobile phases for several labs can be filtered and degassed at a central location, sealed with a three valve cap with the internal inlet filters and tubing connections already in place, and delivered to the HPLC stations ready-for-use.
Furthermore, after an HPLC run is completed, all three valves on each reservoir can be closed to maintain the mobile phase in a sealed environment until the next run. Tetrahydrofuran (THF), which is widely used as a mobile phase for GPC, attacks the Viton® O-rings of the standard CHROMWARE® systems. The integrated and three valve cap systems have been made into THF-resistant systems by replacing all Viton® O-rings with Kalrez® O-rings, and replacing the standard stainless steel inlet filters with CTFE components, with conventional all-stainless-steel filters.
NOTE: To filter HPLC mobile phases directly into the reservoir, use filtration/degassing cap (BP-1915-047) and a mobile phase pickup adapter (BP-1904-048). Each each sold separately.
Ordering information: Complete three valve cap THF resistant reservoir systems consist of:
BP-1901 TORNADO® HPLC Reservoir with Red UV Safety Coating (qty 1)
BP-1903-647 Three Valve Delivery Cap, THF Resistant (qty 1)
BP-2310-118 TORNADO® PTFE Stirrer (qty 1)
BP-1821-018 PEEK 1/4” -28 Nut for 1/8” O.D. Tubing (qty 8)
BP-1822-018 ETFE Ferrule for 1/8” O.D. Tubing (qty 8)
BP-1823-018 PTFE 1/8” O.D. Tubing 10’ (qty 1)
BP-1918-018 ETFE Adapter, 1/4”-28 to 1/8” I.D. Tubing (qty 2)
BP-1916-621 10 micron S.S. Inlet Filter (qty 2)