SP Wilmad-LabGlass Chromatography Cap Adapter, SP Industries
LG-4558-206 LG-4558-200 LG-4558-100 LG-4558-204 LG-4558-108 LG-4558-104 LG-4558-102 LG-4558-202
14008-120EA 431.09 USD
14008-120 14008-118 14008-108 14008-104 14008-116 14008-106 14008-122 14008-112
SP Wilmad-LabGlass Chromatography Cap Adapter, SP Industries
Designed for use on top of LG-4552 chromatography column sections, these borosilicate glass cap adapters are connected with clamps (LG-4661).
Ordering information: Clamps sold separately.