SP Wilmad-LabGlass Mallory Direct Reading Settlometer, SP Industries
Supplier: SP Industries
LG-5601-100
14204-334EA 494.05 USD
14204-334
SP Wilmad-LabGlass Mallory Direct Reading Settlometer, SP Industries
For use in the oxidized sludge process as required for treatment of sewage and other wastes.
  • Graduated in cc per liter and hundredths of a foot
This settlometer is a 2000 mL heavy wall borosilicate glass cylinder.