    Sort by:
  • Search Within Results
BML-111
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  Enzo Life Sciences
Description:   Potent lipoxin A₄ agonist, equiactive as LXA₄ in inhibition of LTB₄ induced PMNL chemotaxis (IC₅₀ = 5 nM).