Would you like to log in again?
Accessories
Supplier: Electron Microscopy Sciences
AMF-EXTW4-US AMF-4300 AMEP-4624 AMEP-4625 AMEFC-4300-US AMEP-4622 AMEP-4623 AMF-EXTW2-US
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
FL 2-Year Extended Warranty AMF-EXTW2-US 103306-584 Each Retrieving Restricted
Customize EVOS FL Base, US, 100 – 240 V - includes On-Board Computer, Integrated Imaging Software, Color Camera, Two Vessel Holders, Brightfield and Phase Contrast, Adjustable Intensity LED, No Objectives, No Light Cubes AMEFC-4300-US 103307-226 Each Retrieving
FL 4-Year Extended Warranty AMF-EXTW4-US 103307-214 Each Retrieving Restricted
Customize EVOS FL base - includes On-Board Computer, Integrated Imaging Software, Brightfield and Phase Contrast, Adjustable Intensity LED, No Objectives, No Light Cubes AMF-4300 103307-224 Each Retrieving
Magnification: 10x, Plan: LWD - FL, Imaging Medium: Air, NA: 0.30; WD: 8.3 mm AMEP-4623 103306-940 Each Retrieving
Magnification: 20x, Plan: LWD - FL, Imaging Medium: Air, NA: 0.45; WD: 7.1 mm AMEP-4624 103307-098 Each Retrieving
Magnification: 40x, Plan: LWD - FL, Imaging Medium: Air, NA: 0.65; WD: 2.8 mm AMEP-4625 103307-144 Each Retrieving
Magnification: 4x, Plan: LWD - FL, Imaging Medium: Air, NA: 0.13; WD: 19.7 mm AMEP-4622 103306-276 Each Retrieving
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty