    Sort by:
  • Search Within Results
BMPO
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Enzo Life Sciences
Description:   Nitrone spin trap for the specific in vivo or in vitro detection of short-lived superoxide, hydroxyl and thiyl radicals.