SP Wilmad-LabGlass Class A Automatic Burettes, SP Industries
Supplier: SP Industries
LG-4261T-110 LG-4261T-112 LG-4261T-114 LG-4261T-134
14205-810EA 1561.76 USD
14205-810 14208-132 14205-850 14205-464
SP Wilmad-LabGlass Class A Automatic Burettes, SP Industries
Made of borosilicate glass, these automatic burettes feature a special three-way, 2mm bore stopcock with PTFE plug through which they may be emptied or filled with provisions for return of overflow liquid to reserve bottle.
They are designed with a relief air valve which automatically closes when air pressure is applied above the liquid in the reservoir.
Supplied with drying tubes at air inlet and at top vent, as well as required rubber components (LG-4340-100). Bottle (LG-3460) sold separately.