DL-Sulforaphane ≥97%

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: 1-Isothiocyanato-4-(methylsulphinyl)butane1-Isothiocyanato-4-methylsulphinyl)butane1-Isothiocyanato-4-methylsulfinyl)butane

0219378125
IC0219378125EA 613.15 USD
IC0219378125
DL-Sulforaphane ≥97%
DL-Sulforaphane

Sulforaphane is an anti-cancer, anti-microbial and anti-diabetic compound found in cruciferous vegetables.
Selective inducer of phase II detoxification enzymes with anticarcinogenic properties. Organosulfur compound found in cruciferous vegetables, including broccoli.

It induces the production of detoxifying enzymes such as quinone reductase and glutathione S-transferase that cause xenobiotic transformation. Sulforaphane also increases the transcription of tumor suppressor proteins and inhibits histone deacetylases. It modulates inflammatory responses by suppressing the LPS-mediated expression of iNOS, COX-2, IL-1β and TNF-α.


Sulforaphane and its glutathione conjugate induced UDP-glucuronosyl transferase (UGT1A1) and glutathione transferase (GSTA1) in cultured cells.

Formula: C₆H₁₁NOS₂
MW: 177.3 g/mol
MDL Number: MFCD00198068
CAS Number: 4478-93-7

Order Now

Specification Test Results

Physical Appearance Liquid
Solubility Soluble in methanol, DMSO (40 mg/mL), ethyl acetate and choloroform. Insoluble in water.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR