SP Wilmad-LabGlass Class A Automatic Burettes with Three-Way Stopcocks, SP Industries
Supplier: SP Industries
LG-4271T-134 LG-4271T-136 LG-4271T-116 LG-4271T-114 LG-4271T-112
14201-060EA 1590.02 USD
14201-060 14200-694 14200-796 14200-544 14201-114
SP Wilmad-LabGlass Class A Automatic Burettes with Three-Way Stopcocks, SP Industries
Made of borosilicate glass, these automatic burets feature a special three-way, 2mm bore stopcock with PTFE plug.
Standard Taper joints fit into reagent bottles with Standard Taper necks. Supplied with required rubber components (LG-4340-100). Bottle (LG-3460) sold separately.