Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SiliaBond® TMA Chloride, SAX, Functionalized Irregular Silica Gel, SiliCycle

Supplier: SILICYCLE INC.
SiliaBond®
R66530B-250G R66530B-100G R66530B-1KG R66630B-1KG R66630B-50G R66630B-100G R66530B-50G R66630B-250G R66630B-5G R66630B-25G R66530B-25G R66530B-5G R66630B-500G R66630B-10G R66530B-10G R66530B-500G
SiliaBond is SiliCycle's line of functionalized silica gels for use in organic synthesis.

  • Great variety of functional groups available
  • Fast Kinetics
  • Compatible with all solvents
  • Excellent Mechanical and Thermal Stability
  • Controlled Loading
  • Available in multiple formats

SiliCycle is a leader in the development of functionalized silica gels, gathered under the name SiliaBond, for use in organic synthesis.

The backbone of all SiliaBond products is SiliCycle SiliaFlash® F60 which provides superior performance for all types of applications thanks to its narrow particle size distribution and high purity.

Supported reagents have distinct advantages over their solution-phase counterparts, the biggest one being the ability to do multiple transformations in a single pot, the immobilization of toxic reagents, and increased selectivity. They are suitable for use in batch reactions and in flow through applications.



 

ORDER

Expand All / Collapse All

TMA Chloride (SAX)
Size Sorbent Particle Size Pore Size Supplier No. VWR Catalog Number Unit Price Quantity
5 g TMA Chloride (SAX), Endcapped 40 - 63 µm 60 Å R66630B-5G 75992-332 Each (5g) Retrieving Restricted
10 g TMA Chloride (SAX), Endcapped 40 - 63 µm 60 Å R66630B-10G 75990-996 Each (10g) Retrieving Restricted
25 g TMA Chloride (SAX), Endcapped 40 - 63 µm 60 Å R66630B-25G 75992-326 Each (25g) Retrieving Restricted
50 g TMA Chloride (SAX), Endcapped 40 - 63 µm 60 Å R66630B-50G 75992-330 Each (50g) Retrieving Restricted
100 g TMA Chloride (SAX), Endcapped 40 - 63 µm 60 Å R66630B-100G 75990-994 Each (100g) Retrieving Restricted
250 g TMA Chloride (SAX), Endcapped 40 - 63 µm 60 Å R66630B-250G 75991-000 Each (250g) Retrieving Restricted
500 g TMA Chloride (SAX), Endcapped 40 - 63 µm 60 Å R66630B-500G 75992-328 Each (500g) Retrieving Restricted
1 kg TMA Chloride (SAX), Endcapped 40 - 63 µm 60 Å R66630B-1KG 75990-998 Each (1kg) Retrieving Restricted

TMA Chloride (SAX), Non-endcapped
Size Sorbent Particle Size Pore Size Supplier No. VWR Catalog Number Unit Price Quantity
5 g TMA Chloride (SAX), Non-endcapped 40 - 63 µm 60 Å R66530B-5G 75990-992 Each (5g) Retrieving Restricted
10 g TMA Chloride (SAX), Non-endcapped 40 - 63 µm 60 Å R66530B-10G 75990-980 Each (10g) Retrieving Restricted
25 g TMA Chloride (SAX), Non-endcapped 40 - 63 µm 60 Å R66530B-25G 75990-986 Each (25g) Retrieving Restricted
50 g TMA Chloride (SAX), Non-endcapped 40 - 63 µm 60 Å R66530B-50G 75990-990 Each (50g) Retrieving Restricted
100 g TMA Chloride (SAX), Non-endcapped 40 - 63 µm 60 Å R66530B-100G 75990-978 Each (100g) Retrieving Restricted
250 g TMA Chloride (SAX), Non-endcapped 40 - 63 µm 60 Å R66530B-250G 75990-984 Each (250g) Retrieving Restricted
500 g TMA Chloride (SAX), Non-endcapped 40 - 63 µm 60 Å R66530B-500G 75990-988 Each (500g) Retrieving Restricted
1 kg TMA Chloride (SAX), Non-endcapped 40 - 63 µm 60 Å R66530B-1KG 75990-982 Each (1kg) Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty