Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Automatic Pinchcock Burettes, Wilmad-LabGlass

Supplier: WILMAD/LABGLASS
LG-4230-110 LG-4230-100 LG-4230-112 LG-4230-102 LG-4230-124 LG-4230-114 LG-4230-104 LG-4230-120
Made of borosilicate glass, these inexpensive portable methods of titration are filled by pressure through a tube extending from the bottle up inside the graduated tube.

A small hole in the filling tube at the zero line permits settling of the liquid level and return of the excess to the bottle.

Furnished complete with burette, bottle, vent tube (LG-4230-130), pinchcock clamp (LG-4647-100), bulb (LG-4340-100), and rubber stopper.

Ordering information: Supplied with #9 Rubber stopper (LG-10392-126) for 10mL, and #10-1/2 rubber stopper (LG-10392-130) for 25 and 50mL.



 

ORDER

Expand All / Collapse All

Complete Automatic Pinchcock Burettes
Capacity Bottle Size Subdivision Supplier No. VWR Catalog Number Unit Price Quantity
10 mL 16 oz. 1/10 mL LG-4230-100 14203-380 Each Retrieving
25 mL 32 oz. 1/10 mL LG-4230-102 14204-514 Each Retrieving
50 mL 32 oz. 1/10 mL LG-4230-104 14209-456 Each Retrieving

Burette Only
Capacity Subdivision Supplier No. VWR Catalog Number Unit Price Quantity
10 mL 1/10 mL LG-4230-110 14204-288 Each Retrieving
25 mL 1/10 mL LG-4230-112 14207-746 Each Retrieving
50 mL 1/10 mL LG-4230-114 14207-236 Each Retrieving

Bottle Only
Bottle Size Subdivision Supplier No. VWR Catalog Number Unit Price Quantity
16 oz. LG-4230-120 14211-514 Each Retrieving
32 oz. LG-4230-124 14201-596 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty