Permanent Polyester Labels, BMP®61 BMP®71 ToughBond®, Brady
Supplier: Brady
BMP®61
148121 148119 148115 148120
75840-051RL 154.89 USD
75840-051 75840-063 75840-053 75840-045
Permanent Polyester Labels, BMP®61 BMP®71 ToughBond®, Brady
Labels
Thermal transfer printable, glossy permanent polyester (B-422) labels are ideal for use in all commercial and industrial applications.
  • Aggressive adhesive is ideal for powder-coated and textured flat surfaces, including low surface energy plastics
  • Designed to withstand exposure to numerous solvents
  • Service temperature of –40 to 100°C (–40 to 212 °F)
  • Colored write-on area
Certifications: ASTM B 117. ASTM D 1000. ASTM D 2979. ASTM G 155. RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC. UL approved. CSA approved.