SiliaBond® C8, Functionalized Irregular Silica Gel, SiliCycle
Supplier: SILICYCLE INC.
SiliaBond®
R31030B-500G R31030B-50G R31030B-500G R31030B-25G R31130B-10G R31130B-500G R31130B-500G R31130B-250G R31130B-1KG R31130B-5G R31030B-5G R31030B-250G R31030B-25G R31130B-10G R31030B-250G R31030B-5G R31030B-10G R31130B-25G R31130B-1KG R31130B-100G R31030B-100G R31130B-50G R31030B-10G R31030B-50G R31030B-100G R31130B-250G R31030B-1KG R31130B-5G R31130B-50G R31130B-25G R31030B-1KG R31130B-100G
75991-172EA 1025.9 USD
75991-172 75991-174 CA75991-172 CA75991-170 75991-180 75991-188 CA75991-188 CA75991-184 75991-182 CA75991-192 75991-176 75991-168 75991-170 CA75991-180 CA75991-168 CA75991-176 CA75991-164 75991-186 CA75991-182 75991-178 CA75991-162 CA75991-190 75991-164 CA75991-174 75991-162 75991-184 75991-166 75991-192 75991-190 CA75991-186 CA75991-166 CA75991-178
SiliaBond® C8, Functionalized Irregular Silica Gel, SiliCycle
Chromatography Sorbents Flash Chromatography Sorbents
SiliaBond is SiliCycle's line of functionalized silica gels for use in organic synthesis.
  • Great variety of functional groups available
  • Fast Kinetics
  • Compatible with all solvents
  • Excellent Mechanical and Thermal Stability
  • Controlled Loading
  • Available in multiple formats
SiliCycle is a leader in the development of functionalized silica gels, gathered under the name SiliaBond, for use in organic synthesis.
The backbone of all SiliaBond products is SiliCycle SiliaFlash® F60 which provides superior performance for all types of applications thanks to its narrow particle size distribution and high purity.
Supported reagents have distinct advantages over their solution-phase counterparts, the biggest one being the ability to do multiple transformations in a single pot, the immobilization of toxic reagents, and increased selectivity. They are suitable for use in batch reactions and in flow through applications.