SiliaBond® C18, 23%, Functionalized Irregular Silica Gel, SiliCycle
Supplier: SILICYCLE INC.
SiliaBond®
R30030B-5G R30130B-250G R30030B-500G R30130B-25G R30130B-25G R30030B-1KG R30030B-10G R30030B-10G R30030B-50G R30130B-10G R30130B-100G R30030B-1KG R30030B-50G R30030B-100G R30130B-50G R30030B-250G R30030B-250G R30130B-500G R30130B-10G R30130B-5G R30030B-25G R30130B-1KG R30130B-50G R30130B-250G R30030B-500G R30030B-100G R30130B-100G R30130B-1KG R30130B-5G R30030B-5G R30130B-500G R30030B-25G
75991-120EA 23.45 USD
75991-120 75990-608 75991-116 75991-122 CA75991-122 CA75991-110 CA75991-108 75991-108 CA75991-118 CA75990-604 CA75990-602 75991-110 75991-118 CA75991-106 CA75991-126 CA75991-112 75991-112 75991-124 75990-604 CA75991-128 75991-114 75990-606 75991-126 CA75990-608 CA75991-116 75991-106 75990-602 CA75990-606 75991-128 CA75991-120 CA75991-124 CA75991-114
SiliaBond® C18, 23%, Functionalized Irregular Silica Gel, SiliCycle
Chromatography Sorbents Flash Chromatography Sorbents
SiliaBond is SiliCycle's line of functionalized silica gels for use in organic synthesis.
  • Great variety of functional groups available
  • Fast Kinetics
  • Compatible with all solvents
  • Excellent Mechanical and Thermal Stability
  • Controlled Loading
  • Available in multiple formats
SiliCycle is a leader in the development of functionalized silica gels, gathered under the name SiliaBond, for use in organic synthesis.
The backbone of all SiliaBond products is SiliCycle SiliaFlash® F60 which provides superior performance for all types of applications thanks to its narrow particle size distribution and high purity.
Supported reagents have distinct advantages over their solution-phase counterparts, the biggest one being the ability to do multiple transformations in a single pot, the immobilization of toxic reagents, and increased selectivity. They are suitable for use in batch reactions and in flow through applications.