SiliaBond® Propyl Chloride, Functionalized Irregular Silica Gel, SiliCycle
Supplier: SiliCycle
SiliaBond®
R59030B-1KG R59030B-500G R59030B-10G R59030B-50G R59030B-100G R59030B-25G R59030B-50G R59030B-10G R59030B-1KG R59030B-25G R59030B-5G R59030B-5G R59030B-250G R59030B-250G R59030B-500G R59030B-100G
75991-660EA 668.46 USD
CA75991-658 CA75991-668 75991-660 75991-662 CA75991-660 75991-654 75991-664 CA75991-662 75991-656 CA75991-654 75991-666 CA75991-664 75991-658 CA75991-656 CA75991-666 75991-668
SiliaBond® Propyl Chloride, Functionalized Irregular Silica Gel, SiliCycle
Chromatography Sorbents Flash Chromatography Sorbents
SiliaBond is SiliCycle's line of functionalized silica gels for use in organic synthesis.
  • Great variety of functional groups available
  • Fast Kinetics
  • Compatible with all solvents
  • Excellent Mechanical and Thermal Stability
  • Controlled Loading
  • Available in multiple formats
SiliCycle is a leader in the development of functionalized silica gels, gathered under the name SiliaBond, for use in organic synthesis.
The backbone of all SiliaBond products is SiliCycle SiliaFlash® F60 which provides superior performance for all types of applications thanks to its narrow particle size distribution and high purity.
Supported reagents have distinct advantages over their solution-phase counterparts, the biggest one being the ability to do multiple transformations in a single pot, the immobilization of toxic reagents, and increased selectivity. They are suitable for use in batch reactions and in flow through applications.