SiliaBond® Diol, 15-40 µm, Functionalized Irregular Silica Gel, SiliCycle
R35017B-1KG R35023B-1KG R35019B-1KG R35017B-1KG R35023B-100G R35019B-1KG R35017B-100G R35019B-100G R35023B-1KG R35017B-100G R35023B-100G R35019B-100G
75991-380EA 2451.46 USD
75991-380 75991-390 75991-382 CA75991-380 CA75991-390 CA75991-382 75991-384 75991-386 CA75991-384 75991-388 CA75991-386 CA75991-388
SiliaBond® Diol, 15-40 µm, Functionalized Irregular Silica Gel, SiliCycle
SiliaBond is SiliCycle's line of functionalized silica gels for use in organic synthesis.
  • Great variety of functional groups available
  • Fast Kinetics
  • Compatible with all solvents
  • Excellent Mechanical and Thermal Stability
  • Controlled Loading
  • Available in multiple formats
SiliCycle is a leader in the development of functionalized silica gels, gathered under the name SiliaBond, for use in organic synthesis.
The backbone of all SiliaBond products is SiliCycle SiliaFlash® F60 which provides superior performance for all types of applications thanks to its narrow particle size distribution and high purity.
Supported reagents have distinct advantages over their solution-phase counterparts, the biggest one being the ability to do multiple transformations in a single pot, the immobilization of toxic reagents, and increased selectivity. They are suitable for use in batch reactions and in flow through applications.