SiliaBond® Cyclohexyl, Functionalized Irregular Silica Gel, SiliCycle
Supplier: SILICYCLE INC.
SiliaBond®
R61530B-500G R61530B-1KG R61530B-50G R61530B-25G R61530B-5G R61530B-50G R61530B-250G R61530B-10G R61530B-500G R61530B-25G R61530B-100G R61530B-1KG R61530B-250G R61530B-100G R61530B-5G R61530B-10G
75991-740EA 1305.17 USD
75991-740 CA75991-734 CA75991-742 75991-738 75992-244 75991-742 75991-736 CA75991-732 CA75991-740 CA75991-738 CA75991-730 75991-734 CA75991-736 75991-730 CA75992-244 75991-732
SiliaBond® Cyclohexyl, Functionalized Irregular Silica Gel, SiliCycle
Chromatography Sorbents Flash Chromatography Sorbents
SiliaBond is SiliCycle's line of functionalized silica gels for use in organic synthesis.
  • Great variety of functional groups available
  • Fast Kinetics
  • Compatible with all solvents
  • Excellent Mechanical and Thermal Stability
  • Controlled Loading
  • Available in multiple formats
SiliCycle is a leader in the development of functionalized silica gels, gathered under the name SiliaBond, for use in organic synthesis.
The backbone of all SiliaBond products is SiliCycle SiliaFlash® F60 which provides superior performance for all types of applications thanks to its narrow particle size distribution and high purity.
Supported reagents have distinct advantages over their solution-phase counterparts, the biggest one being the ability to do multiple transformations in a single pot, the immobilization of toxic reagents, and increased selectivity. They are suitable for use in batch reactions and in flow through applications.