SiliaBond® C18, SPE Grade, Functionalized Irregular Silica Gel, SiliCycle
R31930B-250G R31930B-25G R31930B-5G R31930B-1KG R31930B-10G R31930B-1KG R31930B-50G R31930B-50G R31930B-500G R31930B-100G R31930B-10G R31930B-250G R31930B-100G R31930B-25G R31930B-5G R31930B-500G R31923B-1KG R31923B-1KG
75991-208EA 134.82 USD
75991-208 CA75991-206 CA75991-208 75991-194 75991-196 CA75991-194 75991-210 75991-198 CA75991-196 CA75991-210 75991-200 CA75991-198 CA75991-200 75991-202 CA75991-202 75991-204 CA75991-204 75991-206
SiliaBond® C18, SPE Grade, Functionalized Irregular Silica Gel, SiliCycle
SiliaBond is SiliCycle's line of functionalized silica gels for use in organic synthesis.
  • Great variety of functional groups available
  • Fast Kinetics
  • Compatible with all solvents
  • Excellent Mechanical and Thermal Stability
  • Controlled Loading
  • Available in multiple formats
SiliCycle is a leader in the development of functionalized silica gels, gathered under the name SiliaBond, for use in organic synthesis.
The backbone of all SiliaBond products is SiliCycle SiliaFlash® F60 which provides superior performance for all types of applications thanks to its narrow particle size distribution and high purity.
Supported reagents have distinct advantages over their solution-phase counterparts, the biggest one being the ability to do multiple transformations in a single pot, the immobilization of toxic reagents, and increased selectivity. They are suitable for use in batch reactions and in flow through applications.