SiliaBond® C18, 11% Monomeric, Functionalized Irregular Silica Gel, SiliCycle
Supplier: SILICYCLE INC.
SiliaBond®
R33530B-1KG R33530B-50G R33530B-500G R33530B-100G R33530B-100G R33530B-5G R33530B-10G R33530B-1KG R33530B-25G R33530B-250G R33530B-25G R33530B-5G R33530B-500G R33530B-50G R33530B-250G R33530B-10G
75991-342EA 1337.18 USD
CA75991-336 CA75991-344 75991-342 75991-332 CA75991-332 CA75991-346 75991-334 75991-336 75991-340 CA75991-338 CA75991-340 75991-346 CA75991-342 75991-344 75991-338 CA75991-334
SiliaBond® C18, 11% Monomeric, Functionalized Irregular Silica Gel, SiliCycle
Chromatography Sorbents Flash Chromatography Sorbents
SiliaBond is SiliCycle's line of functionalized silica gels for use in organic synthesis.
  • Great variety of functional groups available
  • Fast Kinetics
  • Compatible with all solvents
  • Excellent Mechanical and Thermal Stability
  • Controlled Loading
  • Available in multiple formats
SiliCycle is a leader in the development of functionalized silica gels, gathered under the name SiliaBond, for use in organic synthesis.
The backbone of all SiliaBond products is SiliCycle SiliaFlash® F60 which provides superior performance for all types of applications thanks to its narrow particle size distribution and high purity.
Supported reagents have distinct advantages over their solution-phase counterparts, the biggest one being the ability to do multiple transformations in a single pot, the immobilization of toxic reagents, and increased selectivity. They are suitable for use in batch reactions and in flow through applications.