Octanoyl-L-Carnitine Chloride
Supplier: INDOFINE CHEMICAL COMPANY MS
17080025MG
100284-506EA
601.54
USD
100284-506
Octanoyl-L-Carnitine Chloride
Octanoyl-L-carnitine chloride
Formula:
C₁₅H₃₀ClNO₄ |
MDL Number:
MFCD00274134 CAS Number: 54377-02-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...