12-Hydroxystearic Acid

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 12-Hydroxyoctadecanoic Acid

141820100MG
100284-398EA 255 USD
100284-398
12-Hydroxystearic Acid
12-Hydroxystearic acid
Formula: C₁₈H₃₆O₃
MW: 300.48 g/mol
MDL Number: MFCD00004592
CAS Number: 106-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR