2-Hydroxy-n-octanoic Acid

Supplier: Avantor

Synonyms: 2-Hydroxy-n-octanoic Acid

14080250MG
100284-174EA 417 USD
100284-174
2-Hydroxy-n-octanoic Acid
2-Hydroxy-n-octanoic acid
Formula: C₈H₁₆O₃
MW: 160.21 g/mol
MDL Number: MFCD00014410
CAS Number: 617-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR