L(-)-Carnitine (from Bovine)

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: R)-(-)-3-Hydroxy-4-(trimethylammonio)butyrateL-CarnitineLevocarnitine(R)-3-Hydroxy-4-(trimethylammonio)butanoateVitamin BT

1700001GM
100284-484EA 657.77 USD
100284-484
L(-)-Carnitine (from Bovine)
L(-)-Carnitine
Formula: C₇H₁₅NO₃
MW: 161.20 g/mol
MDL Number: MFCD00038747
CAS Number: 541-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR