    Sort by:
  • Search Within Results
Plant sterol mixture
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Catalog Number: (100272-526)
Supplier:  Indofine Chemical Company
Description:   Lipid Research 25mg beta-sitosterol 53%, stigmasterol 7%, campesterol 26%, brassicasterol 13% Room temperature.
Minority or Woman-Owned Business Enterprise Small Business Enterprise