Flammable Materials Storage Refrigerators, FMS, Thermo Scientific
Supplier: Thermo Scientific
05FREETSA 20FRCETSA 50FREETSA 20FREETSA
10842-536EA 8325.3 USD InStock
10842-536 10818-362 10031-646 75993-422
Flammable Materials Storage Refrigerators, FMS, Thermo Scientific
Refrigerators
Ensure safe cold storage of volatile materials with Thermo Scientific™ Flammable-Materials Storage Refrigerators
  • Manual defrost
  • Hydraulic thermostat
  • Aluminum evaporator coil
  • Magnetic vinyl gaskets for positive door seal
  • CFC-free insulation and coolant
  • Meets OSHA requirements
These units are engineered with no internal electrical components that could trigger an explosion and meet OSHA and National Fire Protection Association safety requirements. The hydraulic thermostat on the exterior eliminates interaction with stored materials. Compressor is mounted on bottom. Refrigerators have an epoxy-coated steel interior and exterior finish. Insulation is polyurethane foam.
Certifications: Meets OSHA requirements. cCSAus.
Ordering information: Manufacturer's warranty varies by country, please contact your VWR Sales Representative for details.