Econix Silver Nanospheres, Dried, PVP Surface, nanoComposix
Supplier: nanoComposix
SEPD5-500MG SEPD50-500MG
103370-094EA 655 USD
103370-094 103370-088
Econix Silver Nanospheres, Dried, PVP Surface, nanoComposix
Beads Microcarriers
Dried powder silver nanoparticles, redispersible in polar solvents (water, alcohols). Excellent value.
  • High quality nanoparticles, precisely engineered and highly characterized
  • Unagglomerated and monodisperse
  • Dried powder dispersible in a wide variety of solvents
  • Econix material provided in larger quantities and lower cost
Spherical silver nanoparticles with dried PVP excipient to ensure particle stability so the materials redisperse without aggregation
Each batch is extensively characterized to meet highest quality standards, ensuring reliable and reproducible results. All products supplied with batch-specific COA's with TEM imaging, TEM sizing, and UV-Vis.
Colloidal silver has applications in Diagnostics, Antimicrobial coatings, Electronics and Photovoltaics, SERS, Optics, Nanotoxicology, Plasmonics (SPR/Surface Plasmon Resonance), novel Color Enhancements and Pigments, Molecular Detection, Sensors, and Photothermal and Microwave-based Therapies.
Custom formulations and ISO13485/cGMP compliant materials can be provided on request.
Delivery information: Ships at ambient temperatures, via FedEx Priority Overnight (domestic).
Caution: DO NOT FREEZE. Store at 2 to 25 °C in a dry place. Nanomaterials can be sensitive to high and low pH conditions and high salt concentrations. Use caution when changing environmental conditions as particles may aggregate.