Mortar and Pestle, Alumina, Coorstek
Supplier: CoorsTek
60373 60374 60371
70750-002EA 1252.96 USD
70750-002 70750-004 50412-280
Mortar and Pestle, Alumina, Coorstek
Mortars and Pestles Mortars with Pestles
These mortars and pestles are ideal for precise elemental analysis as they provide minimal sample contamination
  • Made of AD-99.5% alumina
  • Extremely hard, non-absorbent, wear-resistant material
  • Octagon-Style Mortar: Glazed completely except inside grinding surface
  • Pestle: Glazed completely except grinding surface