SP Wilmad-LabGlass Air Condensers, SP Industries
Supplier: SP Industries
LG-4670-104
14202-446EA 97.12 USD
14202-446
SP Wilmad-LabGlass Air Condensers, SP Industries
Condensers
These borosilicate glass air condensers consist of a plain 13 mm tube with a Standard Taper 24/40 inner joint with drip tip.