Gibson Assembly® HiFi HC Cloning Master Mixes, SGI-DNA
Supplier: Synthetic Genomics
GA1100-4X10M GA1100-4X50M
75997-588EA 1121.28 USD
75997-588 75997-586
Gibson Assembly® HiFi HC Cloning Master Mixes, SGI-DNA
Gibson Assembly HiFi HC 1-Step Kits and Master Mixes allow restriction digest-free, cloning of one or more DNA fragments into virtually any location of any plasmid vector in a single round of cloning.
  • Fast and efficient - Complex assemblies with > 90% cloning efficiencies in one hour with less than five minutes hands-on time
  • Restriction Digest Free - Clone without the need for restriction enzyme sites
  • Multiple Inserts - Reliable assembly of up to five inserts at one time
  • Optimal for fragment sizes ranging from 500 bp to 32 kb
  • Concentrated 4X mix – Allows for use of more dilute DNA samples
  • Formulated with a thermostable proof-reading polymerase for greater numbers of error-free clones
Assemble seamless, perfect constructs in a single round of cloning using up to five inserts and fragments from 500 bp to 32 kb. Provided as a 4X master mix The Gibson Assembly HiFi HC 1-Step mix enables the cloning of less concentrated DNA samples. With performance identical to the original 2X Gibson Assembly HiFi mix, the Gibson Assembly® HiFi 1-Step method allows you to clone genes into any vector without restriction digestion. Clone directly into your final vector and avoid time consuming subcloning.
Delivery information: Shipped on dry ice. Store at ?20 °C upon arrival.